Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152433.3 | NP_689646.2 | 3 | 1448 | ||
NM_198439.2 | NP_940841.1 | 4 | 1649 | |||
XM_006718767.3 | XP_006718830.1 | 4 | 1679 | |||
XM_011542618.2 | XP_011540920.1 | 3 | 1293 | |||
XM_011542619.2 | XP_011540921.1 | 3 | 1431 | |||
GenBank mRNA | AK055247.1 | - | 1 | 65 | ||
AK092993.1 | - | 3 | 1051 | |||
AK289915.1 | - | 4 | 1259 | |||
BC117466.1 | - | 3 | 1147 | 40126017 | ||
BX640672.1 | - | 3 | 1169 |