Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152641.2 | NP_689854.2 | 9 | 1073 | ||
XM_006719272.3 | XP_006719335.1 | 9 | 1227 | |||
XM_011538025.1 | XP_011536327.1 | 2 | 97 | |||
GenBank mRNA | AK055181.1 | - | 2 | 97 | ||
AL832200.1 | - | 5 | 626 | |||
AY727870.2 | - | 9 | 1073 | |||
BX648152.1 | - | 2 | 281 | |||
CR749833.1 | - | 8 | 992 | |||
DQ096628.1 | - | 9 | 1073 |