Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1D SIRNA Assays ›
Gene Symbol:
EEF1D
Gene Name:
eukaryotic translation elongation factor 1 delta
Gene Aliases:
EF-1D, EF1D, FP1047
Chromosome Location:
Chr.8: 143579697 - 143597675 on Build GRCh38
UniGene ID:
Hs.333388
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4486
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130053.2
NP_001123525.2
4
1502
NM_001130055.2
NP_001123527.1
3
573
NM_001130056.2
NP_001123528.1
2
397
NM_001130057.2
NP_001123529.1
2
579
NM_001195203.1
NP_001182132.1
2
397
NM_001289950.1
NP_001276879.1
2
394
NM_001317743.1
NP_001304672.1
2
394
NM_001960.4
NP_001951.2
2
397
NM_032378.4
NP_115754.3
4
1619
XM_005250820.3
XP_005250877.1
3
6494
XM_005250823.1
XP_005250880.1
3
1228
XM_006716519.3
XP_006716582.1
3
6491
XM_006716520.3
XP_006716583.1
3
6494
XM_006716522.1
XP_006716585.1
3
1482
XM_006716523.1
XP_006716586.1
4
1442
XM_006716524.1
XP_006716587.1
3
1249
XM_006716525.1
XP_006716588.1
2
417
XM_011516905.2
XP_011515207.1
4
6064
XM_011516906.2
XP_011515208.1
6
4656
XM_017013168.1
XP_016868657.1
4
5495
XM_017013169.1
XP_016868658.1
5
5151
XM_017013170.1
XP_016868659.1
4
4937
XM_017013171.1
XP_016868660.1
3
1225
XM_017013172.1
XP_016868661.1
3
1482
XM_017013173.1
XP_016868662.1
2
574
GenBank mRNA
AF370363.1
-
4
1202
AK024550.1
-
4
1356
AK093756.1
-
2
1539
AK225040.1
-
4
1357
AK225243.1
-
2
134
AK225270.1
-
2
135
AK293339.1
-
2
135
AK314311.1
-
4
1238
BC000678.2
-
2
941
BC007847.2
-
4
1223
4129368
BC009907.2
-
1
119
BC012819.2
-
1
119
BC062535.1
-
2
111
BC071840.1
-
2
99
BC094806.1
-
2
108
BG482145.1
-
2
101
BG748894.1
-
2
579
BI667117.1
-
2
402
BQ064320.1
-
254
BT007242.1
-
2
1160
CF128478.1
-
2
484
CN424074.1
-
2
386
CN424083.1
-
2
338
CN424105.1
-
2
182
CX787126.1
-
2
444
Z21507.1
-
2
131
Back To Top
More Information
Panther Classification:
Molecular Function -
translation elongation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to ionizing radiation
cell-cell adhesion
DNA binding
translation elongation factor activity
signal transducer activity
protein binding
translation factor activity, RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0