Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300838.1 | NP_001287767.1 | 6 | 1187 | ||
NM_001300839.1 | NP_001287768.1 | 6 | 933 | |||
NM_001300840.1 | NP_001287769.1 | 6 | 881 | |||
NM_001300841.1 | NP_001287770.1 | 6 | 949 | |||
NM_144697.3 | NP_653298.1 | 5 | 1776 | |||
XM_017000378.1 | XP_016855867.1 | 5 | 1042 | |||
GenBank mRNA | AK056960.1 | - | 5 | 1057 | ||
AK315397.1 | - | 5 | 1461 | |||
BC017397.1 | - | 6 | 905 | |||
BC027999.1 | - | 6 | 1177 | 5194413 | ||
BC047238.1 | - | 5 | 1490 | 5273646 |