Search
Search

GLIS1| Gene Symbol: GLIS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147193.2 | NP_671726.2 | 10 | 2720 | ||
| XM_017000408.1 | XP_016855897.1 | 11 | 2787 | |||
| XM_017000409.1 | XP_016855898.1 | 11 | 2775 | |||
| XM_017000410.1 | XP_016855899.1 | 11 | 2901 | |||
| XM_017000411.1 | XP_016855900.1 | 9 | 2440 | |||
| GenBank mRNA | AK090634.1 | - | 10 | 2719 | ||
| AK093474.1 | - | 8 | 2089 | |||