Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEPT10 SIRNA Assays ›
          Gene Symbol:
          SEPT10 SEPT14
          Gene Name:
          septin 10
          septin 14
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.2: 109542797 - 109614247 on Build GRCh38
          Chromosome Location:
          Chr.7: 55793544 - 55862789 on Build GRCh38
          UniGene ID:
          Hs.469615
          Hs.453629
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s45507

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEPT10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321496.1 NP_001308425.1 3 495
          NM_001321498.1 NP_001308427.1 5 809
          NM_001321500.1 NP_001308429.1 4 740
          NM_001321501.1 NP_001308430.1 5 813
          NM_001321502.1 NP_001308431.1 4 740
          NM_001321504.1 NP_001308433.1 2 426
          NM_001321505.1 NP_001308434.1 2 426
          NM_001321507.1 NP_001308436.1 3 495
          NM_001321508.1 NP_001308437.1 2 426
          NM_001321509.1 NP_001308438.1 5 813
          NM_001321512.1 NP_001308441.1 5 809
          NM_001321513.1 NP_001308442.1 5 946
          NM_001321514.1 NP_001308443.1 5 637
          NM_001321515.1 NP_001308444.1 4 877
          NM_144710.4 NP_653311.1 5 809
          NM_178584.3 NP_848699.1 4 740
          XM_006712317.1 XP_006712380.1 5 809
          XM_011510698.2 XP_011509000.1 5 642
          XM_011510699.2 XP_011509001.1 4 461
          XM_011510700.2 XP_011509002.1 5 812
          XM_011510701.2 XP_011509003.1 3 536
          XM_011510703.2 XP_011509005.1 5 419
          XM_011510704.2 XP_011509006.1 5 809
          GenBank mRNA AF146760.1 - 5 556
          AK021681.1 - 5 809
          AK022682.1 - 2 141
          AK092033.1 - 4 460
          AK292038.1 - 5 527
          AK295560.1 - 3 205
          BC020502.1 - 5 503 3913356
          BC050345.1 - 5 746 4830029
          Gene Symbol: SEPT14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_207366.2 NP_997249.2 5 505
          XM_011515373.2 XP_011513675.1 5 489
          GenBank mRNA AK301928.1 - 5 468
          DC400351.1 - 5 505

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          cell division
          protein binding
          GTP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0