Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040202.1 | NP_001035292.1 | 4 | 762 | ||
XM_005262769.4 | XP_005262826.1 | 4 | 762 | |||
XM_017007785.1 | XP_016863274.1 | 4 | 762 | |||
GenBank mRNA | AK055774.1 | - | 4 | 550 | ||
AK291232.1 | - | 4 | 742 | |||
AX775925.1 | - | 4 | 756 | |||
AY424281.1 | - | 4 | 689 | |||
BC029604.1 | - | 3 | 530 | |||
BC031256.1 | - | 4 | 728 | 5271599 | ||
BC047510.1 | - | 4 | 737 | 5297045 | ||
BX648346.1 | - | 3 | 393 |