Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 11 | 1808 | |||
GenBank mRNA | BC063682.1 | - | 7 | 1154 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 11 | 1806 | ||
XM_011517658.2 | XP_011515960.1 | 11 | 1923 | |||
XM_011517659.2 | XP_011515961.1 | 11 | 1705 | |||
XM_011517660.2 | XP_011515962.1 | 11 | 1702 | |||
XM_011517662.2 | XP_011515964.1 | 11 | 3209 | |||
XM_011517663.2 | XP_011515965.1 | 10 | 3058 | |||
XM_011517664.2 | XP_011515966.1 | 10 | 3055 | |||
XM_011517665.2 | XP_011515967.1 | 10 | 1604 | |||
XM_011517666.2 | XP_011515968.1 | 10 | 1603 | |||
XM_017014169.1 | XP_016869658.1 | 11 | 1822 | |||
XM_017014170.1 | XP_016869659.1 | 11 | 1885 | |||
XM_017014171.1 | XP_016869660.1 | 9 | 1497 | |||
XM_017014172.1 | XP_016869661.1 | 9 | 3075 | |||
GenBank mRNA | AK024481.1 | - | 2 | 4214 | ||
BC094860.1 | - | 3 | 847 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 11 | 1934 | |||
GenBank mRNA | AL137714.1 | - | 8 | 3474 | ||
BC073913.1 | - | 6 | 1323 | |||
BC082240.1 | - | 7 | 2769 | |||
BC093083.1 | - | 6 | 1336 | |||
BX537637.1 | - | 11 | 1573 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 11 | 2487 | |||
GenBank mRNA | AK056232.1 | - | 11 | 2332 | ||
AK057951.1 | - | 13 | 1932 | |||
AK095183.1 | - | 10 | 1928 | |||
AK097269.1 | - | 3 | 1713 | |||
AK290087.1 | - | 9 | 1548 | |||
BC063555.1 | - | 10 | 2122 | |||
BC110991.1 | - | 6 | 1096 | |||
BQ937971.1 | - | 1 | 328 |
Gene Symbol: WASH6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029274.1 | - | 2 | 1616 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 11 | 1702 | |||
GenBank mRNA | AK092583.1 | - | 7 | 3139 | ||
BC063470.1 | - | 7 | 1552 | |||
BC094698.1 | - | 8 | 2483 |