Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138492.5 | NP_612501.3 | 3 | 424 | ||
NM_182960.3 | NP_892005.1 | 2 | 221 | |||
NM_205846.2 | NP_995318.1 | 2 | 221 | |||
XM_017009127.1 | XP_016864616.1 | 2 | 221 | |||
XM_017009128.1 | XP_016864617.1 | 2 | 221 | |||
XM_017009129.1 | XP_016864618.1 | 2 | 221 | |||
XM_017009130.1 | XP_016864619.1 | 2 | 221 | |||
XM_017009131.1 | XP_016864620.1 | 2 | 221 | |||
XM_017009132.1 | XP_016864621.1 | 2 | 221 | |||
XM_017009133.1 | XP_016864622.1 | 2 | 221 | |||
XM_017009134.1 | XP_016864623.1 | 2 | 221 | |||
XM_017009135.1 | XP_016864624.1 | 2 | 221 | |||
GenBank mRNA | AK095695.1 | - | 2 | 156 | ||
AK125215.1 | - | 2 | 156 | |||
BC012033.1 | - | 3 | 391 | 4476359 | ||
BC028175.1 | - | 138 | ||||
BQ645893.1 | - | 2 | 191 | |||
BU167299.1 | - | 2 | 83 | |||
BU186346.1 | - | 2 | 158 | |||
DB266840.1 | - | 2 | 223 |