Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177965.3 | NP_808880.1 | 5 | 494 | ||
GenBank mRNA | AK055162.1 | - | 5 | 469 | ||
BC036855.2 | - | 5 | 459 | |||
DA473498.1 | - | 5 | 494 | |||
DB205618.1 | - | 5 | 459 | |||
GQ240139.1 | - | 5 | 469 |