Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015447.3 | NP_056262.3 | 11 | 3077 | ||
XM_005263396.3 | XP_005263453.1 | 12 | 3502 | |||
XM_005263397.1 | XP_005263454.1 | 9 | 2603 | |||
XM_011518272.1 | XP_011516574.1 | 11 | 2771 | |||
XM_011518273.2 | XP_011516575.1 | 10 | 2602 | |||
XM_011518274.1 | XP_011516576.1 | 9 | 2632 | |||
XM_017014301.1 | XP_016869790.1 | 11 | 3358 | |||
XM_017014302.1 | XP_016869791.1 | 11 | 3282 | |||
GenBank mRNA | AJ519841.1 | - | 9 | 2602 | ||
BC012778.2 | - | 1 | 1077 | |||
BC130580.1 | - | 11 | 2736 | 40146611 | ||
BC144082.1 | - | 11 | 2736 | 9052602 |