Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELAVL2 SIRNA Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like neuron-specific RNA binding protein 2
Gene Aliases:
HEL-N1, HELN1, HUB
Chromosome Location:
Chr.9: 23690099 - 23850757 on Build GRCh38
UniGene ID:
Hs.166109
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4605
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELAVL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171195.1
NP_001164666.1
2
380
NM_001171197.1
NP_001164668.1
2
393
NM_004432.3
NP_004423.2
2
399
XM_005251393.3
XP_005251450.1
2
537
XM_005251394.3
XP_005251451.1
2
694
XM_005251395.3
XP_005251452.1
2
474
XM_006716734.3
XP_006716797.1
3
786
XM_006716735.3
XP_006716798.1
3
254
XM_006716736.3
XP_006716799.1
2
675
XM_011517774.2
XP_011516076.1
3
800
XM_011517776.2
XP_011516078.1
2
230
XM_011517777.2
XP_011516079.1
3
786
XM_011517778.2
XP_011516080.2
2
323
XM_011517779.2
XP_011516081.1
3
298
XM_011517780.2
XP_011516082.1
3
627
XM_011517783.2
XP_011516085.1
3
660
XM_011517784.2
XP_011516086.1
2
355
XM_011517785.2
XP_011516087.1
2
160
XM_011517786.2
XP_011516088.1
2
534
XM_017014408.1
XP_016869897.1
4
801
XM_017014409.1
XP_016869898.1
3
439
XM_017014410.1
XP_016869899.1
3
583
XM_017014411.1
XP_016869900.1
3
766
XM_017014412.1
XP_016869901.1
4
748
XM_017014413.1
XP_016869902.1
4
747
XM_017014414.1
XP_016869903.1
3
770
XM_017014415.1
XP_016869904.1
2
216
XM_017014416.1
XP_016869905.1
3
289
XM_017014417.1
XP_016869906.1
3
660
XM_017014418.1
XP_016869907.1
2
494
XM_017014419.1
XP_016869908.1
2
385
XM_017014420.1
XP_016869909.1
2
388
XM_017014421.1
XP_016869910.1
2
506
XM_017014422.1
XP_016869911.1
2
675
XM_017014423.1
XP_016869912.1
2
332
XM_017014424.1
XP_016869913.1
2
678
XM_017014425.1
XP_016869914.1
2
486
GenBank mRNA
AB209294.1
-
2
399
BC030692.1
-
2
397
4826082
BC042393.1
-
2
383
Back To Top
More Information
Panther Classification:
Molecular Function -
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
nucleotide binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0