Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELAVL2 SIRNA Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like neuron-specific RNA binding protein 2
Gene Aliases:
HEL-N1, HELN1, HUB
Chromosome Location:
Chr.9: 23690099 - 23850757 on Build GRCh38
UniGene ID:
Hs.166109
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4606
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELAVL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171195.1
NP_001164666.1
6
986
NM_001171197.1
NP_001164668.1
6
999
NM_004432.3
NP_004423.2
7
1044
XM_005251393.3
XP_005251450.1
7
1185
XM_005251394.3
XP_005251451.1
7
1342
XM_005251395.3
XP_005251452.1
7
1122
XM_006716734.3
XP_006716797.1
8
1434
XM_006716735.3
XP_006716798.1
8
902
XM_006716736.3
XP_006716799.1
7
1323
XM_011517774.2
XP_011516076.1
8
1445
XM_011517776.2
XP_011516078.1
6
839
XM_011517777.2
XP_011516079.1
7
1392
XM_011517778.2
XP_011516080.2
7
971
XM_011517779.2
XP_011516081.1
8
946
XM_011517780.2
XP_011516082.1
8
1275
XM_011517783.2
XP_011516085.1
8
1308
XM_011517784.2
XP_011516086.1
7
1003
XM_011517785.2
XP_011516087.1
7
808
XM_011517786.2
XP_011516088.1
7
1182
XM_017014408.1
XP_016869897.1
9
1446
XM_017014409.1
XP_016869898.1
8
1084
XM_017014410.1
XP_016869899.1
8
1228
XM_017014411.1
XP_016869900.1
8
1414
XM_017014412.1
XP_016869901.1
9
1396
XM_017014413.1
XP_016869902.1
9
1392
XM_017014414.1
XP_016869903.1
8
1415
XM_017014415.1
XP_016869904.1
7
861
XM_017014416.1
XP_016869905.1
7
895
XM_017014417.1
XP_016869906.1
8
1305
XM_017014418.1
XP_016869907.1
7
1142
XM_017014419.1
XP_016869908.1
7
1030
XM_017014420.1
XP_016869909.1
7
1033
XM_017014421.1
XP_016869910.1
7
1151
XM_017014422.1
XP_016869911.1
7
1320
XM_017014423.1
XP_016869912.1
7
977
XM_017014424.1
XP_016869913.1
6
1284
XM_017014425.1
XP_016869914.1
6
1092
XM_017014426.1
XP_016869915.1
6
663
GenBank mRNA
AB209294.1
-
7
1044
BC030692.1
-
6
1003
4826082
BC042393.1
-
6
989
Back To Top
More Information
Panther Classification:
Molecular Function -
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
nucleotide binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0