Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178498.3 | NP_848593.2 | 7 | 1250 | ||
XM_006718155.3 | XP_006718218.1 | 7 | 1153 | |||
XM_006718156.3 | XP_006718219.1 | 7 | 1489 | |||
XM_011519920.2 | XP_011518222.1 | 8 | 730 | |||
XM_011519921.2 | XP_011518223.1 | 7 | 1490 | |||
XM_017017244.1 | XP_016872733.1 | 8 | 1176 | |||
GenBank mRNA | AY299482.1 | - | 7 | 959 | ||
BC041454.1 | - | 7 | 719 | |||
BC049207.1 | - | 7 | 1222 | 5188979 | ||
CN370367.1 | - | 3 | 359 |