Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTLL9 SIRNA Assays ›
Gene Symbol:
TTLL9
Gene Name:
tubulin tyrosine ligase like 9
Gene Aliases:
C20orf125
Chromosome Location:
Chr.20: 31870702 - 31944959 on Build GRCh38
UniGene ID:
Hs.712915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s46524
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTLL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008409.2
NP_001008409.1
15
1509
NR_134519.1
16
1657
XM_005260309.2
XP_005260366.1
13
1230
XM_006723723.2
XP_006723786.1
11
1061
XM_006723724.2
XP_006723787.1
15
1509
XM_011528617.1
XP_011526919.1
15
1352
XM_011528618.1
XP_011526920.1
15
1516
XM_011528619.1
XP_011526921.1
16
1414
XM_011528620.2
XP_011526922.1
16
1411
XM_011528621.1
XP_011526923.1
16
1429
XM_011528622.1
XP_011526924.1
16
1404
XM_011528623.2
XP_011526925.1
14
1285
XM_011528624.1
XP_011526926.1
14
1283
XM_011528625.1
XP_011526927.1
14
1265
XM_011528626.1
XP_011526928.1
15
1350
XM_011528627.2
XP_011526929.1
15
1347
XM_011528628.1
XP_011526930.1
14
1275
XM_011528629.1
XP_011526931.1
14
1283
XM_011528630.1
XP_011526932.1
13
1217
XM_011528631.1
XP_011526933.1
13
1196
XM_011528632.1
XP_011526934.1
15
1352
XM_011528633.1
XP_011526935.1
15
1390
XM_011528634.1
XP_011526936.1
17
1582
XM_011528635.1
XP_011526937.1
16
1472
XM_011528640.1
XP_011526942.1
13
1068
XM_011528644.1
XP_011526946.1
13
1128
XM_011528646.1
XP_011526948.1
13
1188
XM_017027694.1
XP_016883183.1
12
1148
XM_017027695.1
XP_016883184.1
16
1477
XM_017027696.1
XP_016883185.1
12
980
XM_017027697.1
XP_016883186.1
17
1559
XM_017027698.1
XP_016883187.1
16
1495
XM_017027699.1
XP_016883188.1
15
1655
XM_017027700.1
XP_016883189.1
16
1494
XM_017027701.1
XP_016883190.1
15
1275
XM_017027702.1
XP_016883191.1
15
1407
XM_017027703.1
XP_016883192.1
14
1206
GenBank mRNA
AK090839.1
-
1
336
AK093491.1
-
15
1510
AK123328.1
-
2
1807
AK302360.1
-
11
1063
AK302806.1
-
13
1213
BC068593.1
-
16
1647
BC104024.1
-
13
1188
40025306
BC104025.1
-
15
1509
40025308
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
ATP binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0