Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TTLL9 SIRNA Assays ›
          Gene Symbol:
          TTLL9
          Gene Name:
          tubulin tyrosine ligase like 9
          Gene Aliases:
          C20orf125
          Chromosome Location:
          Chr.20: 31870702 - 31944959 on Build GRCh38
          UniGene ID:
          Hs.712915
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s46525

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TTLL9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008409.2 NP_001008409.1 3 353
          NR_134519.1 4 439
          XM_005260309.2 XP_005260366.1 2 117
          XM_006723724.2 XP_006723787.1 2 226
          XM_011528617.1 XP_011526919.1 3 189
          XM_011528618.1 XP_011526920.1 3 353
          XM_011528619.1 XP_011526921.1 4 251
          XM_011528620.2 XP_011526922.1 4 248
          XM_011528621.1 XP_011526923.1 4 266
          XM_011528622.1 XP_011526924.1 4 241
          XM_011528623.2 XP_011526925.1 2 122
          XM_011528624.1 XP_011526926.1 3 189
          XM_011528625.1 XP_011526927.1 3 189
          XM_011528626.1 XP_011526928.1 3 187
          XM_011528627.2 XP_011526929.1 3 184
          XM_011528628.1 XP_011526930.1 2 112
          XM_011528629.1 XP_011526931.1 2 120
          XM_011528631.1 XP_011526933.1 3 189
          XM_011528632.1 XP_011526934.1 3 189
          XM_011528633.1 XP_011526935.1 3 189
          XM_011528634.1 XP_011526936.1 3 189
          XM_011528635.1 XP_011526937.1 3 189
          XM_011528637.2 XP_011526939.1 3 189
          XM_011528638.1 XP_011526940.1 3 189
          XM_011528639.1 XP_011526941.1 3 189
          XM_011528640.1 XP_011526942.1 3 189
          XM_011528641.1 XP_011526943.1 3 189
          XM_011528643.2 XP_011526945.1 3 189
          XM_011528644.1 XP_011526946.1 3 189
          XM_011528646.1 XP_011526948.1 4 378
          XM_017027695.1 XP_016883184.1 3 189
          XM_017027696.1 XP_016883185.1 3 189
          XM_017027697.1 XP_016883186.1 3 189
          XM_017027698.1 XP_016883187.1 3 189
          XM_017027699.1 XP_016883188.1 2 372
          XM_017027700.1 XP_016883189.1 3 189
          XM_017027701.1 XP_016883190.1 3 189
          XM_017027702.1 XP_016883191.1 3 189
          XM_017027703.1 XP_016883192.1 3 189
          GenBank mRNA AK093491.1 - 3 354
          AK302806.1 - 2 100
          AK303963.1 - 2 102
          BC068593.1 - 4 430
          BC104024.1 - 4 378 40025306
          BC104025.1 - 2 226 40025308

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein modification process
          ATP binding
          ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0