Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTLL9 SIRNA Assays ›
Gene Symbol:
TTLL9
Gene Name:
tubulin tyrosine ligase like 9
Gene Aliases:
C20orf125
Chromosome Location:
Chr.20: 31870702 - 31944959 on Build GRCh38
UniGene ID:
Hs.712915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s46526
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTLL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008409.2
NP_001008409.1
4
385
NR_134519.1
5
471
XM_005260309.2
XP_005260366.1
3
149
XM_006723723.2
XP_006723786.1
2
86
XM_006723724.2
XP_006723787.1
3
258
XM_011528617.1
XP_011526919.1
4
221
XM_011528618.1
XP_011526920.1
4
385
XM_011528619.1
XP_011526921.1
5
283
XM_011528620.2
XP_011526922.1
5
280
XM_011528621.1
XP_011526923.1
5
298
XM_011528622.1
XP_011526924.1
5
273
XM_011528623.2
XP_011526925.1
3
154
XM_011528624.1
XP_011526926.1
4
221
XM_011528625.1
XP_011526927.1
4
221
XM_011528626.1
XP_011526928.1
4
219
XM_011528627.2
XP_011526929.1
4
216
XM_011528628.1
XP_011526930.1
3
144
XM_011528629.1
XP_011526931.1
3
152
XM_011528630.1
XP_011526932.1
2
86
XM_011528631.1
XP_011526933.1
4
221
XM_011528632.1
XP_011526934.1
4
221
XM_011528633.1
XP_011526935.1
4
221
XM_011528634.1
XP_011526936.1
4
221
XM_011528635.1
XP_011526937.1
4
221
XM_011528637.2
XP_011526939.1
4
221
XM_011528638.1
XP_011526940.1
4
221
XM_011528639.1
XP_011526941.1
4
221
XM_011528640.1
XP_011526942.1
4
221
XM_011528641.1
XP_011526943.1
4
221
XM_011528643.2
XP_011526945.1
4
221
XM_011528644.1
XP_011526946.1
4
221
XM_011528646.1
XP_011526948.1
5
410
XM_017027694.1
XP_016883183.1
2
86
XM_017027695.1
XP_016883184.1
4
221
XM_017027696.1
XP_016883185.1
4
221
XM_017027697.1
XP_016883186.1
4
221
XM_017027698.1
XP_016883187.1
4
221
XM_017027699.1
XP_016883188.1
3
404
XM_017027700.1
XP_016883189.1
4
221
XM_017027701.1
XP_016883190.1
4
221
XM_017027702.1
XP_016883191.1
4
221
XM_017027703.1
XP_016883192.1
4
221
GenBank mRNA
AK093491.1
-
4
386
AK302360.1
-
2
88
AK302806.1
-
3
132
AK303963.1
-
3
134
BC068593.1
-
5
462
BC104024.1
-
5
410
40025306
BC104025.1
-
3
258
40025308
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
ATP binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0