siRNA Details
Gene Symbol: ADAMTS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139056.3 | NP_620687.2 | 14 | 2270 | ||
GenBank mRNA | AB095949.1 | - | 11 | 1525 | ||
AJ315734.1 | - | 14 | 2132 | |||
AK127746.1 | - | 14 | 2270 |

More Information
Other Species Targeted

Panther Classification:
