Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152641.2 | NP_689854.2 | 15 | 2728 | ||
XM_006719272.3 | XP_006719335.1 | 15 | 2882 | |||
XM_011538025.1 | XP_011536327.1 | 9 | 1835 | |||
GenBank mRNA | AL832200.1 | - | 11 | 2281 | ||
AY727870.2 | - | 15 | 2728 | |||
BX537879.1 | - | 1 | 356 | |||
BX648152.1 | - | 9 | 2016 | |||
BX649133.1 | - | 7 | 1574 | |||
CR749833.1 | - | 14 | 2647 | |||
DQ096628.1 | - | 15 | 2728 |