Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105513.2 | NP_001098983.2 | 24 | 3487 | ||
NM_145755.2 | NP_665698.2 | 25 | 3631 | |||
XM_005264921.4 | XP_005264978.1 | 25 | 3602 | |||
XM_005264922.4 | XP_005264979.1 | 25 | 3587 | |||
XM_005264923.3 | XP_005264980.1 | 25 | 3560 | |||
XM_005264924.4 | XP_005264981.1 | 24 | 3491 | |||
XM_005264925.4 | XP_005264982.1 | 24 | 3479 | |||
XM_005264926.4 | XP_005264983.1 | 22 | 3248 | |||
XM_005264927.4 | XP_005264984.1 | 21 | 3056 | |||
XM_006713011.3 | XP_006713074.1 | 25 | 3575 | |||
XM_011533447.2 | XP_011531749.1 | 24 | 3503 | |||
XM_017005843.1 | XP_016861332.1 | 22 | 3206 | |||
GenBank mRNA | AJ487015.1 | - | 25 | 3593 | ||
AK093313.1 | - | 15 | 2171 | |||
AK098528.1 | - | 7 | 997 | |||
AK302408.1 | - | 24 | 3470 | |||
AK302465.1 | - | 24 | 3341 | |||
BX647142.1 | - | 6 | 1203 |