Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182532.2 | NP_872338.1 | 2 | 455 | ||
XM_005270586.4 | XP_005270643.1 | 2 | 93 | |||
XM_011540911.2 | XP_011539213.1 | 3 | 207 | |||
XM_011540912.2 | XP_011539214.1 | 2 | 983 | |||
GenBank mRNA | BC029775.1 | - | 2 | 327 | 5186268 | |
BG430102.1 | - | 2 | 455 |