Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIKFYVE SIRNA Assays ›
Gene Symbol:
PIKFYVE
Gene Name:
phosphoinositide kinase, FYVE-type zinc finger containing
Gene Aliases:
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
Chromosome Location:
Chr.2: 208266178 - 208358751 on Build GRCh38
UniGene ID:
Hs.744997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47255
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIKFYVE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178000.1
NP_001171471.1
7
1056
NM_015040.3
NP_055855.2
7
1056
NM_152671.3
NP_689884.1
5
765
XM_011510778.2
XP_011509080.1
8
1106
XM_011510779.2
XP_011509081.1
8
1039
XM_011510780.2
XP_011509082.1
8
1124
XM_011510781.2
XP_011509083.1
8
1065
XM_011510782.2
XP_011509084.1
8
1106
XM_011510783.2
XP_011509085.1
8
1106
XM_011510784.2
XP_011509086.1
8
1124
XM_011510785.2
XP_011509087.1
8
1064
XM_011510786.2
XP_011509088.1
6
773
XM_011510787.1
XP_011509089.1
6
782
XM_011510788.1
XP_011509090.1
5
765
XM_011510789.2
XP_011509091.1
4
587
XM_011510790.1
XP_011509092.1
6
884
XM_011510792.2
XP_011509094.1
8
1106
XM_017003568.1
XP_016859057.1
7
960
XM_017003569.1
XP_016859058.1
7
960
XM_017003570.1
XP_016859059.1
5
791
XM_017003571.1
XP_016859060.1
4
587
XM_017003572.1
XP_016859061.1
6
760
XM_017003573.1
XP_016859062.1
3
468
XM_017003574.1
XP_016859063.1
2
139
GenBank mRNA
AK127769.1
-
7
960
AY457063.1
-
7
1060
BC017736.2
-
6
884
BC032389.1
-
5
765
BC125052.1
-
6
787
BC125053.1
-
7
1045
EU794609.1
-
7
1060
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
receptor-mediated endocytosis
myelin assembly
protein localization to nucleus
intracellular signal transduction
phosphatidylinositol-3-phosphate biosynthetic process
retrograde transport, endosome to Golgi
phosphatidylinositol phosphorylation
phosphatidylinositol 5-phosphate metabolic process
regulation of autophagosome assembly
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
zinc ion binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0