Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIKFYVE SIRNA Assays ›
Gene Symbol:
PIKFYVE
Gene Name:
phosphoinositide kinase, FYVE-type zinc finger containing
Gene Aliases:
CFD, FAB1, HEL37, PIP5K, PIP5K3, ZFYVE29
Chromosome Location:
Chr.2: 208266178 - 208358751 on Build GRCh38
UniGene ID:
Hs.744997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47256
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIKFYVE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178000.1
NP_001171471.1
9
1286
NM_015040.3
NP_055855.2
9
1286
NM_152671.3
NP_689884.1
7
995
XM_011510778.2
XP_011509080.1
10
1336
XM_011510779.2
XP_011509081.1
10
1269
XM_011510780.2
XP_011509082.1
10
1354
XM_011510781.2
XP_011509083.1
10
1295
XM_011510782.2
XP_011509084.1
10
1336
XM_011510783.2
XP_011509085.1
10
1336
XM_011510784.2
XP_011509086.1
10
1354
XM_011510785.2
XP_011509087.1
10
1294
XM_011510786.2
XP_011509088.1
8
1003
XM_011510787.1
XP_011509089.1
8
1012
XM_011510788.1
XP_011509090.1
7
995
XM_011510789.2
XP_011509091.1
6
817
XM_011510790.1
XP_011509092.1
8
1114
XM_011510792.2
XP_011509094.1
10
1336
XM_017003568.1
XP_016859057.1
9
1190
XM_017003569.1
XP_016859058.1
9
1190
XM_017003570.1
XP_016859059.1
7
1021
XM_017003571.1
XP_016859060.1
6
817
XM_017003572.1
XP_016859061.1
8
990
XM_017003573.1
XP_016859062.1
5
698
XM_017003574.1
XP_016859063.1
4
369
GenBank mRNA
AK127769.1
-
9
1191
AY457063.1
-
9
1290
BC017736.2
-
8
1114
BC032389.1
-
7
995
BC125052.1
-
8
1017
BC125053.1
-
9
1275
EU794609.1
-
9
1290
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
receptor-mediated endocytosis
myelin assembly
protein localization to nucleus
intracellular signal transduction
phosphatidylinositol-3-phosphate biosynthetic process
retrograde transport, endosome to Golgi
phosphatidylinositol phosphorylation
phosphatidylinositol 5-phosphate metabolic process
regulation of autophagosome assembly
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
zinc ion binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-v95b6:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0