Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYSND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040273.2 | NP_001035363.1 | 2 | 1992 | ||
NM_173555.3 | NP_775826.2 | 4 | 2309 | |||
NR_073580.1 | 2 | 760 | ||||
NR_073581.1 | 4 | 1197 | ||||
NR_073582.1 | 4 | 1077 | ||||
NR_073591.1 | 4 | 988 | ||||
NR_073592.1 | 2 | 880 | ||||
NR_073593.1 | 4 | 980 | ||||
NR_073594.1 | 4 | 1100 | ||||
NR_073595.1 | 4 | 1233 | ||||
GenBank mRNA | AK074968.1 | - | 4 | 1135 | ||
AK097697.1 | - | 3 | 1852 | |||
BC016840.1 | - | 4 | 1763 | |||
BC030242.1 | - | 2 | 1908 | 5176098 | ||
BC047424.1 | - | 2 | 760 | |||
BC111501.1 | - | 4 | 2248 | 5732356 |