Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCNY SIRNA Assays ›
Gene Symbol:
CCNY
Gene Name:
cyclin Y
Gene Aliases:
C10orf9, CBCP1, CCNX, CFP1
Chromosome Location:
Chr.10: 35246981 - 35572670 on Build GRCh38
UniGene ID:
Hs.14745
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47720
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCNY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282852.1
NP_001269781.1
3, 4
478
NM_001282853.1
NP_001269782.1
5, 6
616
NM_145012.5
NP_659449.3
4, 5
553
NM_181698.3
NP_859049.2
6, 7
577
XM_006717392.3
XP_006717455.1
4, 5
322
XM_011519358.1
XP_011517660.1
7, 8
644
XM_011519360.2
XP_011517662.1
8, 9
746
XM_011519361.1
XP_011517663.1
8, 9
628
XM_011519362.1
XP_011517664.1
7, 8
608
XM_011519363.1
XP_011517665.1
7, 8
621
XM_011519364.1
XP_011517666.1
9, 10
742
XM_011519365.2
XP_011517667.1
8, 9
661
XM_011519366.1
XP_011517668.1
7, 8
591
XM_011519367.1
XP_011517669.1
5, 6
421
XM_011519368.2
XP_011517670.1
4, 5
316
XM_017015839.1
XP_016871328.1
4, 5
239
XM_017015840.1
XP_016871329.1
4, 5
241
XM_017015841.1
XP_016871330.1
4, 5
239
GenBank mRNA
AF413522.1
-
4, 5
553
AF429969.1
-
6, 7
533
AF465728.1
-
4, 5
299
AF465729.1
-
4, 5
374
AK057280.1
-
6, 7
533
AK314666.1
-
5, 6
592
AL834355.1
-
4, 5
411
AY504868.1
-
4, 5
553
BC069224.1
-
4, 5
379
BC094815.1
-
4, 5
405
6340750
BC104773.1
-
5, 6
362
8143776
BC104801.1
-
5, 6
362
8143804
BC143450.1
-
4, 5
255
9051958
BC143455.1
-
5, 6
362
9051963
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
Wnt signaling pathway
positive regulation of cyclin-dependent protein serine/threonine kinase activity
cell division
regulation of canonical Wnt signaling pathway
protein binding
cyclin-dependent protein serine/threonine kinase regulator activity
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0