Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR9Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005212.3 | NP_001005212.1 | 3 | 371 | ||
GenBank mRNA | BC110819.1 | - | 3 | 371 | 5203967 |