Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM21C SIRNA Assays ›
Gene Symbol:
FAM21C
Gene Name:
family with sequence similarity 21 member C
Gene Aliases:
FAM21A, VPEF
Chromosome Location:
Chr.10: 45727170 - 45792964 on Build GRCh38
UniGene ID:
Hs.365286
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s48438
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM21C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001169106.1
NP_001162577.1
9
899
NM_001169107.1
NP_001162578.1
9
899
NM_015262.2
NP_056077.2
9
899
XM_011539566.1
XP_011537868.1
9
866
XM_011539567.1
XP_011537869.1
9
866
XM_011539568.1
XP_011537870.1
9
866
XM_011539569.1
XP_011537871.1
9
866
XM_011539570.2
XP_011537872.1
8
755
XM_011539571.1
XP_011537873.1
9
866
XM_011539572.1
XP_011537874.1
9
866
XM_017016014.1
XP_016871503.1
9
866
XM_017016015.1
XP_016871504.1
9
866
XM_017016016.1
XP_016871505.1
9
866
XM_017016017.1
XP_016871506.1
9
866
XM_017016018.1
XP_016871507.1
9
866
XM_017016019.1
XP_016871508.1
9
866
XM_017016020.1
XP_016871509.1
9
866
XM_017016021.1
XP_016871510.1
9
794
XM_017016022.1
XP_016871511.1
9
840
XM_017016023.1
XP_016871512.1
9
784
XM_017016026.1
XP_016871515.1
9
866
GenBank mRNA
AB011164.1
-
9
880
AK291430.1
-
9
887
AK293929.1
-
7
613
AK298385.1
-
7
564
AK303048.1
-
9
863
BC053887.1
-
8
651
BM126842.1
-
5
354
BQ229875.1
-
8
689
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
retrograde transport, endosome to Golgi
regulation of substrate adhesion-dependent cell spreading
retrograde transport, endosome to plasma membrane
negative regulation of barbed-end actin filament capping
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
phosphatidylinositol-5-phosphate binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-3,4-bisphosphate binding
phosphatidylinositol-4-phosphate binding
phosphatidylinositol-3,5-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0