Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MECOM SIRNA Assays ›
Gene Symbol:
MECOM
Gene Name:
MDS1 and EVI1 complex locus
Gene Aliases:
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location:
Chr.3: 169083499 - 169663781 on Build GRCh38
UniGene ID:
Hs.744090
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4874
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MECOM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105077.3
NP_001098547.3
13
2654
NM_001105078.3
NP_001098548.2
12
2645
NM_001163999.1
NP_001157471.1
11
2565
NM_001164000.1
NP_001157472.1
11
3494
NM_001205194.1
NP_001192123.1
12
2949
NM_004991.3
NP_004982.2
13
3287
NM_005241.3
NP_005232.2
12
2589
XM_005247213.3
XP_005247270.1
13
3296
XM_005247214.3
XP_005247271.1
12
3269
XM_005247215.3
XP_005247272.1
12
3265
XM_005247219.2
XP_005247276.1
12
3261
XM_005247220.2
XP_005247277.1
12
2602
XM_005247221.2
XP_005247278.1
12
2585
XM_005247223.2
XP_005247280.1
12
3258
XM_005247224.3
XP_005247281.1
13
2324
XM_005247225.3
XP_005247282.1
13
2320
XM_005247226.3
XP_005247283.1
12
2171
XM_011512546.2
XP_011510848.1
12
3049
XM_017005874.1
XP_016861363.1
12
3964
XM_017005875.1
XP_016861364.1
11
2572
XM_017005876.1
XP_016861365.1
11
3234
XM_017005877.1
XP_016861366.1
12
2296
XM_017005878.1
XP_016861367.1
11
2259
GenBank mRNA
AK025934.1
-
6
493
AK292865.1
-
12
2572
AK310509.1
-
10
2406
BC031019.1
-
5
750
BC130520.1
-
13
2628
40146551
BC143951.1
-
13
2628
9052469
BC143952.1
-
11
2539
9052470
BX640908.1
-
11
3494
BX647613.1
-
11
3226
GQ352634.1
-
12
2853
X54989.1
-
12
2587
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
apoptotic process
cell differentiation
histone lysine methylation
negative regulation of programmed cell death
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of JNK cascade
regulation of cell cycle
hematopoietic stem cell proliferation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
histone-lysine N-methyltransferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0