Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TIPRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031800.2 | NP_001026970.1 | 2 | 442 | ||
NM_152902.4 | NP_690866.1 | 2 | 442 | |||
GenBank mRNA | AB097034.1 | - | 2 | 325 | ||
AK313520.1 | - | 2 | 442 | |||
AL049670.1 | - | 2 | 354 | |||
BC009506.2 | - | 2 | 232 | 3532344 |