Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172139.2 | NP_742151.2 | 1 | 130 | ||
XM_005258765.4 | XP_005258822.1 | 2 | 542 | |||
GenBank mRNA | AY129149.1 | - | 1 | 138 | ||
AY184374.1 | - | 1 | 126 | |||
AY336714.1 | - | 1 | 129 | |||
AY336717.1 | - | 1 | 129 | |||
BC130314.1 | - | 1 | 165 | 40146345 | ||
BC130316.1 | - | 1 | 165 | 40146347 |
Gene Symbol: IFNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172138.1 | NP_742150.1 | 2 | 190 | ||
GenBank mRNA | AY129148.1 | - | 2 | 190 | ||
AY184373.1 | - | 1 | 126 | |||
AY336715.1 | - | 1 | 129 | |||
BC113581.1 | - | 2 | 193 | 8322633 | ||
BC113583.1 | - | 2 | 193 | 8322635 | ||
DQ126337.1 | - | 2 | 190 |