Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005244.4 | NP_005235.3 | 10 | 1296 | ||
NM_172110.3 | NP_742108.2 | 10 | 1296 | |||
XM_005260327.2 | XP_005260384.1 | 8 | 743 | |||
GenBank mRNA | AF055015.1 | - | 6 | 610 | ||
AK293032.1 | - | 10 | 1000 | |||
BC000289.2 | - | 4 | 448 | |||
BC008803.2 | - | 10 | 978 | 3944547 | ||
BC013882.2 | - | 10 | 1296 | |||
BT006682.1 | - | 9 | 850 | |||
U71207.1 | - | 9 | 924 | |||
U81601.1 | - | 1 | 40 |