Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173598.4 | NP_775869.3 | 10, 11 | 1663 | ||
XM_011538224.2 | XP_011536526.1 | 10, 11 | 1992 | |||
XM_011538225.2 | XP_011536527.1 | 10, 11 | 2457 | |||
XM_011538226.2 | XP_011536528.1 | 10, 11 | 1998 | |||
XM_011538228.2 | XP_011536530.1 | 8, 9 | 822 | |||
XM_011538229.2 | XP_011536531.1 | 10, 11 | 1998 | |||
XM_017019208.1 | XP_016874697.1 | 10, 11 | 1998 | |||
XM_017019209.1 | XP_016874698.1 | 10, 11 | 1998 | |||
XM_017019210.1 | XP_016874699.1 | 5, 6 | 448 | |||
GenBank mRNA | AK098831.1 | - | 8, 9 | 841 | ||
AY345972.1 | - | 8, 9 | 1332 | |||
BC107106.1 | - | 8, 9 | 1327 | |||
BC107107.1 | - | 8, 9 | 1327 | |||
BC127603.1 | - | 8, 9 | 1332 | 40019272 |