Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037331.3 | NP_001032408.1 | 2 | 290 | ||
NM_001303482.1 | NP_001290411.1 | 2 | 290 | |||
XM_011525928.1 | XP_011524230.1 | 4 | 6600 | |||
GenBank mRNA | BC037871.2 | - | 2 | 269 | ||
BC107486.1 | - | 2 | 221 | 40032395 |