Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122962.1 | NP_001116434.1 | 3 | 988 | ||
NM_001134836.1 | NP_001128308.1 | 3 | 694 | |||
XM_005260708.4 | XP_005260765.1 | 3 | 797 | |||
XM_005260709.3 | XP_005260766.1 | 3 | 660 | |||
XM_011529221.2 | XP_011527523.1 | 3 | 1584 | |||
XM_011529223.2 | XP_011527525.1 | 4 | 1032 | |||
XM_011529224.2 | XP_011527526.1 | 3 | 476 | |||
XM_011529225.2 | XP_011527527.1 | 2 | 397 | |||
GenBank mRNA | AK095499.1 | - | 3 | 797 | ||
AK294440.1 | - | 4 | 683 | |||
AK297072.1 | - | 3 | 588 | |||
AY424277.1 | - | 3 | 466 | |||
BM285394.1 | - | 3 | 989 | |||
BQ635335.1 | - | 3 | 1056 |