Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173821.2 | NP_776182.2 | 2 | 1156 | ||
XM_011510995.2 | XP_011509297.1 | 2 | 2520 | |||
XM_011510997.2 | XP_011509299.1 | 2 | 1161 | |||
GenBank mRNA | AK090909.1 | - | 2 | 1161 | ||
BC113517.1 | - | 2 | 1156 | 8322569 | ||
BC113519.1 | - | 2 | 1156 | 8322571 |