Search
Search

ACSL4| Gene Symbol: ACSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318509.1 | NP_001305438.1 | 8 | 1527 | ||
| NM_001318510.1 | NP_001305439.1 | 8 | 1236 | |||
| NM_004458.2 | NP_004449.1 | 8 | 1222 | |||
| NM_022977.2 | NP_075266.1 | 9 | 1534 | |||
| XM_005262109.2 | XP_005262166.1 | 7 | 1396 | |||
| XM_006724635.2 | XP_006724698.1 | 7 | 1014 | |||
| XM_011530888.2 | XP_011529190.1 | 8 | 1276 | |||
| XM_011530889.2 | XP_011529191.1 | 7 | 1282 | |||
| GenBank mRNA | AB061713.1 | - | 8 | 1116 | ||
| AB061714.1 | - | 8 | 1076 | |||
| AF030555.1 | - | 8 | 1223 | |||
| AK292070.1 | - | 8 | 1118 | |||
| AK294915.1 | - | 8 | 1206 | |||
| AK307566.1 | - | 8 | 1385 | |||
| BC034959.2 | - | 8 | 1085 | 4824121 | ||
| Y12777.1 | - | 8 | 1086 | |||