Search
Search

ERVK13-1| Gene Symbol: ERVK13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040023.1 | 3 | 3361 | |||
| GenBank mRNA | AK091794.1 | - | 3 | 1153 | ||
| AK095440.1 | - | 1 | 958 | |||
| AK097014.1 | - | 2 | 893 | |||
| AK131509.1 | - | 1 | 1987 | |||
| AX747190.1 | - | 3 | 1153 | |||
| BC043252.1 | - | 3 | 3363 | |||