Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HYAL1 SIRNA Assays ›
          Gene Symbol:
          HYAL1
          Gene Name:
          hyaluronoglucosaminidase 1
          Gene Aliases:
          HYAL-1, LUCA1, MPS9, NAT6
          Chromosome Location:
          Chr.3: 50299889 - 50312951 on Build GRCh38
          UniGene ID:
          Hs.75619
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s501327

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HYAL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033159.3 NP_149349.2 4 1179
          NM_153281.1 NP_695013.1 6 1446
          NM_153282.2 NP_695014.1 3 1089
          NM_153283.2 NP_695015.1 4 822
          NM_153285.2 NP_695017.1 4 404
          NR_047690.1 3 1664
          XM_011533667.2 XP_011531969.1 5 1569
          XM_011533668.2 XP_011531970.1 4 1460
          XM_011533669.2 XP_011531971.1 5 1857
          GenBank mRNA AF118821.1 - 4 1144
          AF173154.1 - 4 1150
          AF502904.1 - 3 1061
          AF502905.1 - 4 794
          AF502906.1 - 4 814
          AF502907.1 - 4 376
          AF502908.1 - 3 227
          AK097287.1 - 6 1446
          AK290816.1 - 4 1171
          BC025774.1 - 3 1314
          BC035695.1 - 4 1133 5760636
          CR541933.1 - 2 929
          KU178142.1 - 3 1019
          KU178143.1 - 2 929
          U03056.2 - 3 1635
          U96078.1 - 3 1629

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          carbohydrate metabolic process
          inflammatory response
          response to virus
          positive regulation of epithelial cell migration
          chondroitin sulfate catabolic process
          hyaluronan metabolic process
          hyaluronan biosynthetic process
          hyaluronan catabolic process
          positive regulation of cell growth
          negative regulation of cell growth
          cellular response to platelet-derived growth factor stimulus
          cellular response to fibroblast growth factor stimulus
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of growth
          response to antibiotic
          positive regulation of epithelial cell proliferation
          cartilage development
          embryonic skeletal joint morphogenesis
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to pH
          cellular response to UV-B
          positive regulation of hyaluranon cable assembly
          virus receptor activity
          hyalurononglucosaminidase activity
          transcription factor binding
          hyaluronan synthase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0