Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POTEB2 SIRNA Assays ›
          Gene Symbol:
          POTEB2 LOC100288966 LOC102723502 POTEB POTEB3 POTEC POTED
          Gene Name:
          POTE ankyrin domain family member B2
          POTE ankyrin domain family member D-like
          POTE ankyrin domain family member B-like
          POTE ankyrin domain family member B
          POTE ankyrin domain family member B3
          POTE ankyrin domain family member C
          POTE ankyrin domain family member D
          Gene Aliases:
          -
          -
          -
          A26B1, CT104.5, POTE-15, POTE15
          POTE-15, POTEB
          A26B2, CT104.6, POTE-18, POTE18
          A26B3, ANKRD21, CT104.1, POTE, POTE-21, POTE21
          Chromosome Location:
          Chr.15: 20835372 - 20866428 on Build GRCh38
          Chromosome Location:
          Chr.Un|NT_167208: 108007 - 139435 on Build GRCh38
          Chromosome Location:
          Chr.15|NT_187382: 132942 - 167347 on Build GRCh38
          Chromosome Location:
          Chr.15: 21846329 - 21877393 on Build GRCh38
          Chromosome Location:
          Chr.15: 21408964 - 21440429 on Build GRCh38
          Chromosome Location:
          Chr.18: 14507339 - 14543600 on Build GRCh38
          Chromosome Location:
          Chr.21: 13610081 - 13641585 on Build GRCh38
          UniGene ID:
          Hs.722659
          Hs.442712
          -
          Hs.722659
          Hs.722659
          Hs.454726
          Hs.742365
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s50238

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POTEB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277303.1 NP_001264232.1 10 1406
          NR_102390.1 8 1197
          GenBank mRNA BC101007.1 - 8 1197 40006489
          Gene Symbol: LOC100288966
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257362.1 NP_001244291.1 10 1536
          GenBank mRNA AY172978.1 - 10 1536
          Gene Symbol: LOC102723502
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006724900.3 XP_006724963.1 10 1631
          Gene Symbol: POTEB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277304.1 NP_001264233.1 10 1406
          NR_102391.1 8 1197
          Gene Symbol: POTEB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_207355.3 NP_997238.2 10 1855
          GenBank mRNA AY465170.1 - 11 1425
          AY465173.1 - 10 1532
          BC101005.1 - 10 1406 40006483
          BC101006.1 - 10 1517 40006485
          BC127123.1 - 10 1517 40006490
          Gene Symbol: POTEC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001137671.1 NP_001131143.1 10 1938
          NR_134878.1 9 1867
          GenBank mRNA AK307658.1 - 10 1938
          BX649118.1 - 9 1853
          Gene Symbol: POTED
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_174981.3 NP_778146.2 10 1536

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0