Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DYSF SIRNA Assays ›
Gene Symbol:
DYSF
Gene Name:
dysferlin
Gene Aliases:
FER1L1, LGMD2B, MMD1
Chromosome Location:
Chr.2: 71453155 - 71686763 on Build GRCh38
UniGene ID:
Hs.252180
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s502426
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DYSF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130455.1
NP_001123927.1
2, 3
297
NM_001130976.1
NP_001124448.1
2, 3
529
NM_001130977.1
NP_001124449.1
2, 3
529
NM_001130978.1
NP_001124450.1
2, 3
529
NM_001130979.1
NP_001124451.1
2, 3
529
NM_001130980.1
NP_001124452.1
2, 3
529
NM_001130981.1
NP_001124453.1
2, 3
529
NM_001130982.1
NP_001124454.1
2, 3
297
NM_001130983.1
NP_001124455.1
2, 3
297
NM_001130984.1
NP_001124456.1
2, 3
297
NM_001130985.1
NP_001124457.1
2, 3
297
NM_001130986.1
NP_001124458.1
2, 3
297
NM_001130987.1
NP_001124459.1
2, 3
297
NM_003494.3
NP_003485.1
2, 3
529
XM_005264584.4
XP_005264641.1
2, 3
312
XM_005264585.4
XP_005264642.1
2, 3
997
GenBank mRNA
AF075575.1
-
2, 3
526
BC171803.1
-
2, 3
502
DQ267935.1
-
2, 3
297
EU515155.1
-
2, 3
430
EU515156.1
-
2, 3
430
EU515157.1
-
2, 3
430
EU515158.1
-
2, 3
430
EU515159.1
-
2, 3
430
EU515160.1
-
2, 3
430
EU515161.1
-
2, 3
297
EU515162.1
-
2, 3
297
EU515163.1
-
2, 3
297
EU515164.1
-
2, 3
297
EU515165.1
-
2, 3
297
EU515166.1
-
2, 3
297
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
plasma membrane repair
vesicle fusion
muscle contraction
calcium ion binding
protein binding
phospholipid binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0