Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DYSF SIRNA Assays ›
          Gene Symbol:
          DYSF
          Gene Name:
          dysferlin
          Gene Aliases:
          FER1L1, LGMD2B, MMD1
          Chromosome Location:
          Chr.2: 71453155 - 71686763 on Build GRCh38
          UniGene ID:
          Hs.252180
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s502427

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DYSF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130455.1 NP_001123927.1 31, 32 3583
          NM_001130976.1 NP_001124448.1 30, 31 3773
          NM_001130977.1 NP_001124449.1 30, 31 3773
          NM_001130978.1 NP_001124450.1 31, 32 3815
          NM_001130979.1 NP_001124451.1 32, 33 3908
          NM_001130980.1 NP_001124452.1 31, 32 3866
          NM_001130981.1 NP_001124453.1 31, 32 3866
          NM_001130982.1 NP_001124454.1 32, 33 3676
          NM_001130983.1 NP_001124455.1 31, 32 3583
          NM_001130984.1 NP_001124456.1 30, 31 3541
          NM_001130985.1 NP_001124457.1 31, 32 3634
          NM_001130986.1 NP_001124458.1 30, 31 3541
          NM_001130987.1 NP_001124459.1 31, 32 3634
          NM_003494.3 NP_003485.1 31, 32 3815
          XM_005264584.4 XP_005264641.1 32, 33 3691
          XM_005264585.4 XP_005264642.1 32, 33 4376
          GenBank mRNA AF075575.1 - 31, 32 3812
          AK074104.1 - 17, 18 2085
          AK295004.1 - 2, 3 272
          BC171803.1 - 31, 32 3788
          DQ267935.1 - 31, 32 3583
          EU515155.1 - 32, 33 3809
          EU515156.1 - 30, 31 3674
          EU515157.1 - 31, 32 3716
          EU515158.1 - 31, 32 3767
          EU515159.1 - 30, 31 3674
          EU515160.1 - 31, 32 3767
          EU515161.1 - 32, 33 3676
          EU515162.1 - 30, 31 3541
          EU515163.1 - 31, 32 3583
          EU515164.1 - 31, 32 3634
          EU515165.1 - 30, 31 3541
          EU515166.1 - 31, 32 3634

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane repair
          vesicle fusion
          muscle contraction
          calcium ion binding
          protein binding
          phospholipid binding
          calcium-dependent phospholipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0