Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLDN16 SIRNA Assays ›
          Gene Symbol:
          CLDN16
          Gene Name:
          claudin 16
          Gene Aliases:
          HOMG3, PCLN1
          Chromosome Location:
          Chr.3: 190387872 - 190412143 on Build GRCh38
          UniGene ID:
          Hs.251391
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s502790

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CLDN16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006580.3 NP_006571.1 2 649
          GenBank mRNA AF152101.1 - 2 469
          AK314955.1 - 2 648
          BC069662.1 - 2 469 7262563
          BC069682.1 - 2 469 7262480
          BC069759.1 - 2 469 7262492
          BC069777.1 - 2 469 7262504
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA621288.1 - 2 368 Chr. 3 - 190388154 - 190404931 [+] on Build GRCh38
          GenBankmRNA BG433757.1 - 314 Chr. - - [-] on Build
          GenBankmRNA CD622944.1 - 2 343 Chr. 3 - 190388177 - 190404860 [+] on Build GRCh38
          GenBankmRNA DA636744.1 - 2 359 Chr. 3 - 190388163 - 190404874 [+] on Build GRCh38
          GenBankmRNA CD622942.1 - 2 342 Chr. 3 - 190388179 - 190404860 [+] on Build GRCh38
          GenBankmRNA DA539435.1 - 2 282 Chr. 3 - 190388239 - 190408418 [+] on Build GRCh38
          GenBankmRNA DA623344.1 - 2 370 Chr. 3 - 190388152 - 190404904 [+] on Build GRCh38
          GenBankmRNA BP274478.1 - 2 367 Chr. 3 - 190388154 - 190408335 [+] on Build GRCh38
          GenBankmRNA CD622948.1 - 2 315 Chr. 3 - 190388210 - 190404860 [+] on Build GRCh38
          GenBankmRNA CD622947.1 - 2 293 Chr. 3 - 190388227 - 190404860 [+] on Build GRCh38
          GenBankmRNA BP276177.1 - 2 271 Chr. 3 - 190388250 - 190408382 [+] on Build GRCh38
          GenBankmRNA EL736324.1 - 2 401 Chr. 3 - 190388120 - 190408329 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          cellular metal ion homeostasis
          excretion
          calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
          positive regulation of calcium ion transport
          transepithelial transport
          magnesium ion transmembrane transport
          structural molecule activity
          protein binding
          magnesium ion transmembrane transporter activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0