Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012643.3 | NP_001012661.1 | 2, 3 | 581 | ||
XM_017026742.1 | XP_016882231.1 | 1, 2 | 1326 | |||
GenBank mRNA | BC044311.1 | - | 2, 3 | 581 | 5756168 | |
BU196283.1 | - | 473 |