Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178565.4 | NP_848660.3 | 2, 3 | 758 | ||
XM_017013395.1 | XP_016868884.1 | 2, 3 | 747 | |||
GenBank mRNA | AK123023.1 | - | 2, 3 | 721 | ||
AY358166.1 | - | 2, 3 | 411 |