Search
Search

FDPS| Gene Symbol: FDPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135821.1 | NP_001129293.1 | 3 | 369 | ||
| NM_001135822.1 | NP_001129294.1 | 1, 2 | 226 | |||
| NM_001242824.1 | NP_001229753.1 | 2 | 247 | |||
| NM_002004.3 | NP_001995.1 | 3 | 424 | |||
| XM_005244963.1 | XP_005245020.1 | 2 | 192 | |||
| GenBank mRNA | AK021828.1 | - | 1 | 1160 | ||
| AK022841.1 | - | 1 | 1135 | |||
| AK291084.1 | - | 3 | 225 | |||
| BC010004.2 | - | 2 | 213 | 4132071 | ||
| BP370037.1 | - | 2 | 251 | |||
| CN346026.1 | - | 1, 2 | 196 | |||
| D14697.1 | - | 3 | 319 | |||
| DA610405.1 | - | 2 | 192 | |||
| J05262.1 | - | 1 | 13 | |||