Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286509.1 | NP_001273438.1 | 2 | 266 | ||
NM_001286510.1 | NP_001273439.1 | 2 | 265 | |||
NM_001286511.1 | NP_001273440.1 | 2 | 216 | |||
NM_001286512.1 | NP_001273441.1 | 2 | 265 | |||
NM_001286513.1 | NP_001273442.1 | 2 | 266 | |||
NM_001286520.1 | NP_001273449.1 | 2 | 266 | |||
NM_178148.3 | NP_835361.1 | 2 | 266 | |||
GenBank mRNA | AB097021.1 | - | 2 | 223 | ||
AB097039.1 | - | 2 | 223 | |||
AB106538.1 | - | 2 | 114 | |||
AK075456.1 | - | 2 | 195 | |||
AK123749.1 | - | 2 | 223 | |||
AK293344.1 | - | 2 | 223 | |||
AK307572.1 | - | 2 | 201 | |||
AY491520.1 | - | 2 | 232 | |||
BC024288.1 | - | 2 | 230 | 4890949 | ||
CN370928.1 | - | 2 | 166 | |||
DA370522.1 | - | 2 | 265 | |||
DA383304.1 | - | 2 | 241 | |||
DA424392.1 | - | 2 | 223 | |||
DA817588.1 | - | 2 | 216 | |||
DB010939.1 | - | 2 | 201 | |||
HY044970.1 | - | 2 | 267 |