Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C12ORF42 SIRNA Assays ›
Gene Symbol:
C12orf42
Gene Name:
chromosome 12 open reading frame 42
Gene Aliases:
-
Chromosome Location:
Chr.12: 103047669 - 103525258 on Build GRCh38
UniGene ID:
Hs.534649
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s51561
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C12orf42
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099336.2
NP_001092806.1
3, 4
420
NM_001278419.1
NP_001265348.1
3, 4
420
NM_001278420.1
NP_001265349.1
3, 4
285
NM_198521.3
NP_940923.2
3, 4
285
NR_103526.1
3, 4
285
XM_006719377.3
XP_006719440.1
2, 3
223
XM_011538291.1
XP_011536593.1
3, 4
280
XM_011538292.2
XP_011536594.1
3, 4
249
XM_011538293.2
XP_011536595.1
3, 4
256
XM_011538295.1
XP_011536597.1
3, 4
312
XM_011538305.1
XP_011536607.1
3, 4
482
XM_011538306.1
XP_011536608.1
3, 4
280
XM_011538308.2
XP_011536610.1
4, 5
310
XM_011538312.2
XP_011536614.1
4, 5
308
XM_011538315.2
XP_011536617.1
4, 5
301
XM_017019276.1
XP_016874765.1
3, 4
374
XM_017019279.1
XP_016874768.1
6, 7
1054
XM_017019281.1
XP_016874770.1
4, 5
305
GenBank mRNA
AK058052.1
-
3, 4
243
BC039352.1
-
3, 4
243
BC044617.1
-
3, 4
381
5267756
BC117392.1
-
3, 4
243
40125943
BC117394.1
-
3, 4
243
40125945
BI830750.1
-
3, 4
362
HY002043.1
-
3, 4
378
HY015666.1
-
3, 4
285
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0