Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRC37A3 SIRNA Assays ›
Gene Symbol:
LRRC37A3
Gene Name:
leucine rich repeat containing 37 member A3
Gene Aliases:
LRRC37, LRRC37A
Chromosome Location:
Chr.17: 64854147 - 64966228 on Build GRCh38
UniGene ID:
Hs.551962
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s51588
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRC37A3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303255.1
NP_001290184.1
8
668
NM_199340.3
NP_955372.2
12
3925
XM_011524776.1
XP_011523078.1
9
1364
XM_011524777.1
XP_011523079.1
7
1211
XM_011524778.2
XP_011523080.2
10
889
XM_017024593.1
XP_016880082.1
10
3754
XM_017024594.1
XP_016880083.1
9
3682
XM_017024595.1
XP_016880084.1
9
3682
XM_017024596.1
XP_016880085.1
8
3601
XM_017024597.1
XP_016880086.1
8
3601
XM_017024598.1
XP_016880087.1
7
3529
XM_017024599.1
XP_016880088.1
11
3126
XM_017024601.1
XP_016880090.1
9
1066
XM_017024602.1
XP_016880091.1
6
841
XM_017024603.1
XP_016880092.1
9
1364
XM_017024604.1
XP_016880093.1
8
1292
XM_017024605.1
XP_016880094.1
7
1211
XM_017024606.1
XP_016880095.1
6
1139
XM_017024607.1
XP_016880096.1
9
669
XM_017024608.1
XP_016880097.1
9
1686
GenBank mRNA
AB011135.2
-
11
3790
AK091625.1
-
2
773
AK294371.1
-
8
656
AK299714.1
-
5
422
AK308554.1
-
5
401
AK315945.1
-
9
1397
BC113643.1
-
2
287
BC117274.1
-
2
287
BI560889.1
-
9
706
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-wzbw8:80/100.66.73.6:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0