Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002255.1 | NP_001002255.1 | 1 | 250 | ||
GenBank mRNA | AY340238.1 | - | 1 | 250 | ||
BC130305.1 | - | 1 | 244 | 40146336 |