Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007537.2 | NP_001007538.1 | 1 | 69 | ||
NR_104426.1 | 3 | 259 | ||||
XM_011535070.2 | XP_011533372.1 | 2 | 1199 | |||
GenBank mRNA | BC110413.1 | - | 3 | 259 | ||
BC137004.1 | - | 1 | 118 | 9020997 | ||
BC137006.1 | - | 1 | 118 | 9020999 | ||
BC144450.1 | - | 1 | 118 |
Gene Symbol: C1QTNF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303137.1 | NP_001290066.1 | 3 | 194 | ||
NM_001303138.1 | NP_001290067.1 | 2 | 134 | |||
NM_178540.4 | NP_848635.2 | 2 | 110 | |||
GenBank mRNA | AY358145.1 | - | 2 | 105 | ||
BC040438.1 | - | 2 | 110 | 5227612 | ||
BI767787.1 | - | 2 | 132 | |||
BQ882976.1 | - | 201 |